Learn More
Mettler Toledo™ F1AC OIML Class Standard Weight Set (F1AC PL E)
Catalog No. 01912677 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Weight: 1 mg to 500 mg
Catalog No.
01-912-677 Weight: 1 mg to 500 mg
1 options
Catalog No. 01-912-677 Supplier Mettler Toledo™ Supplier No. 30402723
Add to Cart
Add to Cart
F1 OIML class weight set is housed in a plastic box and includes accessories for handling and cleaning.
  • Without calibration certificate
  • For Standard Enviroments: Affordable quality weights for regular routine testing without the need for traceability
TRUSTED_SUSTAINABILITY
Specifications
Product Type Test Weight
Material Stainless Steel
Case Material Plastic
Weight (Metric) 1 to 500 mg
Density 7950 (± 140) kg/m3
Includes 12 Pieces; Without certificate