Learn More
Cayman Chemical F1trIfluoroacetate salt 10mg
SDP
Catalog No. 502912608
Encompass_Preferred
An anti-inflammatory peptide; inhibits LPS-induced phosphorylation of IKKα and IKKβ and nuclear translocation of NF-κB (p65) in isolated mouse peritoneal macrophages at 5 µM; decreases serum levels of IL-6, TNF-α, and IL-1β and increases survival in a mouse model of LPS-induced sepsis at 5 mg/kg
Catalog No. 50-291-2608 Supplier Cayman Chemical Supplier No. 3584610 MG
An anti-inflammatory peptide; inhibits LPS-induced phosphorylation of IKKα and IKKβ and nuclear translocation of NF-κB (p65) in isolated mouse peritoneal macrophages at 5 µM; decreases serum levels of IL-6, TNF-α, and IL-1β and increases survival in a mouse model of LPS-induced sepsis at 5 mg/kg