Learn More
Aviva Systems Biology Corp F2 Antibody - middle region (ARP41757_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41757_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against F2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human F2. Alternate names: PTApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Sheep, Zebrafish
Catalog No. 50-014-57984