Learn More
Kimberly-Clark™ Professional F20 Polycarbonate Face Shields
Catalog No. 19071083
Click to view available options
Lens Material:
Acetate
Catalog No. Lens Material
19-071083 Acetate
1 options
This item is not returnable. View return policy
Catalog No. 19-071083 Supplier Kimberly-Clark™ Professional Supplier No. 29097
This item is not returnable. View return policy
Kimberly-Clark™ Professional F20 Polycarbonate Face Shields are designed for light industrial manufacturing, general splash protection, landscaping and food processing
  • Provide on-the-job protection for heat applications, as well as grinding, flying debris and splashes
  • Made of durable polycarbonate
  • Reusable, depending on the task
  • D3 rating for splash protection
  • Polycarbonate material provides 99.9% UVA/UVB/UVC protection
  • Compatible with Jackson Safety Hard Hat Interchange System
  • Snaps into place on compatible head gear and hard hats – no special tools required
  • When used with any Jackson Safety Brand Headgear, meets the performance standards of ANSI Z87.1+ for high-impact protection and is CSA Z94.3 compliant (when used with compatible Jackson Safety headgear)
Certifications
Meet ANSI Z87.1 2010 Standards when used with any Jackson Safety ™Brand Headgear.
Specifications
Lens Tint Green
Lens Material Acetate