Learn More
Julabo USA Inc Each, F250 recirculating chiller 250W
SDP
Catalog No. 507533945
Encompass_Preferred
Chillers, Julabo, F250 Recirculating Cooler -10 to +40 °C with 250W cooling power. JULABO F Series Recirculating Coolers are a great way to replace tap water and are ideal for basic cooling tasks with 250 W to 1 kW cooling power. Models include a splash-proof membrane keypad with LED temperature display and integrated timer. Easy filling / draining with a fill level indicator and overflow port on the back. Units constructed of stainless steel and high grade plastic with stainless steel internal tank. Centrifugal pressure pump provides circulation to external application.May be used with water, water/glycol mixtures and silicone fluids.The F250 comes with 8 and 10 mm hose barbs. The F500 / F1000 come with 8 and 12 mm hose barbs.Included with each unit: 2 each barbed fittings for tubing 8 and 10 mm inner dia. (pump connections M10x1 female)
Catalog No. 50-753-3945 Supplier Julabo USA Inc Supplier No. 9620025.02
Add to Cart
Add to Cart
Request a Quote
Chillers, Julabo, F250 Recirculating Cooler -10 to +40 °C with 250W cooling power. JULABO F Series Recirculating Coolers are a great way to replace tap water and are ideal for basic cooling tasks with 250 W to 1 kW cooling power. Models include a splash-proof membrane keypad with LED temperature display and integrated timer. Easy filling / draining with a fill level indicator and overflow port on the back. Units constructed of stainless steel and high grade plastic with stainless steel internal tank. Centrifugal pressure pump provides circulation to external application.May be used with water, water/glycol mixtures and silicone fluids.The F250 comes with 8 and 10 mm hose barbs. The F500 / F1000 come with 8 and 12 mm hose barbs.Included with each unit: 2 each barbed fittings for tubing 8 and 10 mm inner dia. (pump connections M10x1 female)