Learn More
Aviva Systems Biology Corp F2R Antibody - C-terminal region (ARP60033_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60033_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against F2R. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human F2R. Alternate names: CF2R, HTR, PAR-1, PAR1, TRApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-924