Learn More
Aviva Systems Biology Corp F2RL2 Antibody - C-terminal region (ARP60037_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60037_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against F2RL2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human F2RL2. Alternate names: PAR3, PAR-3ApplicationWB. Species reactivity: Cow, Human, Pig
Catalog No. 50-286-926