Learn More
STEMCELL Technologies Anti-Mouse F4/80 Antibody, Clone BM8, APC, Size: 25 ug
SDP
Supplier:  STEMCELL Technologies 60027AZ.1
Encompass
Clone BM8 is a rat monoclonal antibody that reacts with the F4/80 antigen (also termed Ly-71 in mouse and EMR-1 in human) an ~160 kDa transmembrane glycoprotein belonging to the EGF-TM7 family of G-protein-coupled receptors. Verified for use with flow cytometry assays use this antibody in applications requiring the detection of human and mouse F4/80. F4/80 is reportedly the only macrophage marker suitable for distinguishing destructive from non-destructive inflammatory processes in the pancreas. The protein is thought to play a role in the generation of CD8 regulatory T cells involved in immune tolerance. This antibody clone has also been verified for purity assessments of cells isolated with EasySep kits including EasySep Mouse Monocyte Enrichment Kit (Catalog 19761) and EasySep Mouse CD11b Positive Selection Kit (Catalog 18770). Visit STEMCELL.com for more information and related products.
Catalog No. 50-239-0414