Learn More
Enzo Life Sciences F4/80 Recombinant monoclonal antibody (Recombinant CI:A3-1) (Rabbit IgG kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6380200
Encompass_Preferred
Clone: Recombinant CI:A3-1. Isotype: IgGκ. Immunogen: Thioglycollate stimulated peritoneal macrophages of mouse origin. Species reactivity: Mouse. Applications: Flow Cytometry, IHC. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3162