Learn More
Bulbman Inc F4T5/BLB BLACKLIGHT BULB
SDP
Catalog No. 501253588
Encompass_Preferred
F4T5/BLB: 4 WATT T5 Black Light Blue Fluorescent G5 Base - 4 Watt T5 Preheat Fluorescent Black Light Blue Bulb, Miniature Bi-Pin (G5) Base. UVA Source Integral Dark Blue Filter.
Catalog No. 50-125-3588 Supplier Bulbman Inc Supplier No. 112379
F4T5/BLB: 4 WATT T5 Black Light Blue Fluorescent G5 Base - 4 Watt T5 Preheat Fluorescent Black Light Blue Bulb, Miniature Bi-Pin (G5) Base. UVA Source Integral Dark Blue Filter.