Learn More
Bulbman Inc F6T5/CW Fluorescent Light Bulb, 6 Watt
SDP
Catalog No. NC9838915
Encompass_Preferred
Fluorescent Light Bulb, 6W, Black Light Tube with Purple Glass, G5 Base, 5/8 in. dia. x 6 in.L, 7500 Life Hours
Catalog No. NC9838915 Supplier Bulbman Inc Supplier No. 107663
Fluorescent Light Bulb, 6W, Black Light Tube with Purple Glass, G5 Base, 5/8 in. dia. x 6 in.L, 7500 Life Hours