Learn More
Aviva Systems Biology Corp F7 Antibody - middle region (ARP60039_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60039_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against F7. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human F7. Alternate names: SPCAApplicationWB. Species reactivity: Cow, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-286-928