Learn More
Aviva Systems Biology Corp F8 Antibody - C-terminal region (ARP60040_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60040_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against F8. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human F8. Alternate names: AHF, DXS1253E, F8B, F8C, FVIII, HEMAApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Rabbit, Sheep
Catalog No. 50-286-929