Learn More
Grainger F8572 Inventory Control Label 3 in H PK1000
Catalog No. 502765241
Encompass_Preferred
The Inventory Control Label from Inventory Control Labels is a 2" x 1" white label with a permanent adhesive backing. It is made from a thermal transfer printable polyester material. The label can withstand temperatures from -40°F to 300°F. It has a shelf life of 1 year when stored at 72°F and 50% relative humidity. The label is smudge proof and water resistant. It can be used for barcoding, inventory control, shipping labels, and more. The label meets the AIAG standard and comes 500 labels per roll.
Catalog No. 50-276-5241 Supplier Grainger Supplier No. 16U840
The Inventory Control Label from Inventory Control Labels is a 2" x 1" white label with a permanent adhesive backing. It is made from a thermal transfer printable polyester material. The label can withstand temperatures from -40°F to 300°F. It has a shelf life of 1 year when stored at 72°F and 50% relative humidity. The label is smudge proof and water resistant. It can be used for barcoding, inventory control, shipping labels, and more. The label meets the AIAG standard and comes 500 labels per roll.