Learn More
Grainger F8572 Inventory Control Label 2 in W PK1000
Catalog No. 502764991
Encompass_Preferred
The Tripp Lite SMART1500LCDT 1500VA 900W UPS Battery Back Up, LCD 120V, Line-Interactive, Tower, 8 Outlets, Tel/DSL, USB, DB9 Serial is an uninterruptible power supply that provides battery backup power and surge protection for electronics and computers. It has a 1500VA/900W power capacity and 8 outlets to connect devices. The LCD display shows power conditions and battery charge level. It provides automatic voltage regulation to correct brownouts and overvoltages without using battery power. The UPS has tel/DSL protection, USB port, DB9 serial port, and includes PowerAlert monitoring software. It has a compact upright tower form factor.
Catalog No. 50-276-4991 Supplier Grainger Supplier No. 16U839
The Tripp Lite SMART1500LCDT 1500VA 900W UPS Battery Back Up, LCD 120V, Line-Interactive, Tower, 8 Outlets, Tel/DSL, USB, DB9 Serial is an uninterruptible power supply that provides battery backup power and surge protection for electronics and computers. It has a 1500VA/900W power capacity and 8 outlets to connect devices. The LCD display shows power conditions and battery charge level. It provides automatic voltage regulation to correct brownouts and overvoltages without using battery power. The UPS has tel/DSL protection, USB port, DB9 serial port, and includes PowerAlert monitoring software. It has a compact upright tower form factor.