Learn More
Grainger F8588 Inventory Control Label Red PK1000
Catalog No. 502767311
Encompass_Preferred
The Grainger 16U821 is a 3M Scotch-Brite General Purpose Hand Pad that can be used for cleaning and scouring. It features an open web construction with non-scratching fibers that are gentle on surfaces. The pad measures 6 inches by 9 inches and has a medium duty abrasiveness. It is ideal for cleaning pots, pans, dishes, counters, appliances, tools, and more. The pad can be used wet or dry and is disposable. It effectively removes grease, dirt, grime, and more from a variety of surfaces while being gentle on finishes. The 3M Scotch-Brite General Purpose Hand Pad from Grainger provides an effective cleaning solution.
Catalog No. 50-276-7311 Supplier Grainger Supplier No. 16U821
The Grainger 16U821 is a 3M Scotch-Brite General Purpose Hand Pad that can be used for cleaning and scouring. It features an open web construction with non-scratching fibers that are gentle on surfaces. The pad measures 6 inches by 9 inches and has a medium duty abrasiveness. It is ideal for cleaning pots, pans, dishes, counters, appliances, tools, and more. The pad can be used wet or dry and is disposable. It effectively removes grease, dirt, grime, and more from a variety of surfaces while being gentle on finishes. The 3M Scotch-Brite General Purpose Hand Pad from Grainger provides an effective cleaning solution.