Learn More
Grainger F8588 Inventory Control Label Green PK1000
Catalog No. 502766077
Encompass_Preferred
The Tripp Lite SmartRack 16U Open Frame Rack is a sturdy, reliable solution for housing EIA-standard 19-inch rack equipment in network closets and back offices. Made from heavy-duty steel with a black powder-coated finish, it features an open frame design for easy access and ventilation. The rack has square mounting holes for easy installation and a weight capacity of 200 lbs. Its overall height is 28 inches and width is 23.75 inches, making it an ideal open rack solution for server rooms or wiring closets.
Catalog No. 50-276-6077 Supplier Grainger Supplier No. 16U820
The Tripp Lite SmartRack 16U Open Frame Rack is a sturdy, reliable solution for housing EIA-standard 19-inch rack equipment in network closets and back offices. Made from heavy-duty steel with a black powder-coated finish, it features an open frame design for easy access and ventilation. The rack has square mounting holes for easy installation and a weight capacity of 200 lbs. Its overall height is 28 inches and width is 23.75 inches, making it an ideal open rack solution for server rooms or wiring closets.