Learn More
Grainger Vikan Hand Scraper, Polypropylene, 4.3 in., Purple, 1 ea
Catalog No. 502777955
Encompass_Preferred
This color-coded hand scraper is designed for efficient removal of dried-on materials and residues in food manufacturing, food retail, restaurants, and food service environments. The one-piece polypropylene construction features no seams or cracks, which effectively prevents bacterial growth and supports stringent hygiene protocols.
  • Beveled scraping edge dislodges burnt-on food, grease, and sticky substances
  • Hygienic one-piece design lacks seams or cracks to harbor bacteria
  • Tapered handle with thumb indentation provides a secure and ergonomic grip
  • Drop-shaped hanging hole facilitates easy storage and prevents pooling liquid
  • Color-coded to support hygienic zoning plans and 5S programs
  • Autoclave safe and made from FDA-compliant materials
  • Operating temperature range of -22°F to 250°F
Catalog No. 50-277-7955 Supplier Grainger Supplier No. 18G807
This color-coded hand scraper is designed for efficient removal of dried-on materials and residues in food manufacturing, food retail, restaurants, and food service environments. The one-piece polypropylene construction features no seams or cracks, which effectively prevents bacterial growth and supports stringent hygiene protocols.
  • Beveled scraping edge dislodges burnt-on food, grease, and sticky substances
  • Hygienic one-piece design lacks seams or cracks to harbor bacteria
  • Tapered handle with thumb indentation provides a secure and ergonomic grip
  • Drop-shaped hanging hole facilitates easy storage and prevents pooling liquid
  • Color-coded to support hygienic zoning plans and 5S programs
  • Autoclave safe and made from FDA-compliant materials
  • Operating temperature range of -22°F to 250°F