Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich F8bt

Catalog No. 698687250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-868-7250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 69-868-7250MG Supplier Sigma Aldrich Supplier No. 698687250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

F8BT is a fluorene copolymer that is used as a light-emitting polymer (LEP) with low energy of lowest unoccupied molecular orbital (LUMO), high electron mobility and luminance. F8BT can be used with poly(9,9-di-n-octylfluorenyl-2,7-diyl) (PTO) to map the effect of noise sources on photoconducting charge transports by evaluating the conductivity and noise source density (Nt). It may also form a nanocomposite with single walled carbon nanotube (SWCNT) with excellent electric properties and high Seebeck coefficient, for potential application in the development of photovoltaic power and biomass energy. Organic light emitting diode (OLED) devices can be fabricated by depositing a blend of F8 and F8BT on PEDOT:PSS films.

Specifications

CAS 210347-52-7
Linear Formula (C35H42N2S)n
Quantity 250 mg
Synonym Poly(9,9-dioctylfluorene-alt-benzothiadiazole); Poly[(9,9-di-n-octylfluorenyl-2,7-diyl)-alt-(benzo[2,1,3 ]thiadiazol-4,8-diyl)]
Molecular Weight (g/mol) 25000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.