Learn More
Biomatik Corporation F9 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC10978
Encompass_Preferred
F9 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to F9. The F9 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Coagulation Factor Ix Protein (144-239Aa). Applications: ELISA, IHC, IF. Target Synonyms: F9. Accession Number: P00740. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7622