Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC F9170 (TFA) | 99.7% | 1983.3 g/mol | 5 MG
SDP

Catalog No. 5000425462
Encompass_Preferred

F9170 (TFA) is a laboratory chemical with 99.7% purity and a molecular weight of 1983.3 g/mol. It appears as a solid and is stable under recommended storage conditions. This product is intended for research use only and should be handled by suitably qualified and experienced scientists in appropriate facilities.

  • Solid appearance
  • Stable under recommended storage conditions
  • Store powder at -80°C for up to 2 years or -20°C for up to 1 year
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month (sealed storage, away from moisture)
  • Suitable for shipping at room temperature if less than 2 weeks
  • Requires handling in areas with appropriate exhaust ventilation
  • Keep container tightly sealed in a cool, well-ventilated area
  • Keep away from direct sunlight and sources of ignition

Catalog No. 50-004-25462 Supplier Medchemexpress LLC Supplier No. HYP10310A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

F9170 (TFA) is a laboratory chemical with 99.7% purity and a molecular weight of 1983.3 g/mol. It appears as a solid and is stable under recommended storage conditions. This product is intended for research use only and should be handled by suitably qualified and experienced scientists in appropriate facilities.

  • Solid appearance
  • Stable under recommended storage conditions
  • Store powder at -80°C for up to 2 years or -20°C for up to 1 year
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month (sealed storage, away from moisture)
  • Suitable for shipping at room temperature if less than 2 weeks
  • Requires handling in areas with appropriate exhaust ventilation
  • Keep container tightly sealed in a cool, well-ventilated area
  • Keep away from direct sunlight and sources of ignition

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.