Learn More
Cayman Chemical F9170trIfluoroacetAT salt 10mg
SDP
Catalog No. 502915460
Encompass_Preferred
An antiviral peptide; binds to the LLP1 domain within the HIV-1 envelope protein cytoplasmic tail; inhibits HIV-1 IIIB infection of MT-2 cells (EC50 = 0.19 µM); reduces plasma viral load in a rhesus macaque model of SHIV infection at 3 mg/kg
Catalog No. 50-291-5460 Supplier Cayman Chemical Supplier No. 3696210 MG
An antiviral peptide; binds to the LLP1 domain within the HIV-1 envelope protein cytoplasmic tail; inhibits HIV-1 IIIB infection of MT-2 cells (EC50 = 0.19 µM); reduces plasma viral load in a rhesus macaque model of SHIV infection at 3 mg/kg