Learn More
ATCC F98; Glioma; Rat (Rattus norvegicus)
Catalog No. 502383592
Encompass
F98 is a glial-like cell that was isolated from the brain of a rat with undifferentiated malignant glioma. F98 can be used as rat brain tumor models in experimental neuro-oncology.
Catalog No. 50-238-3592 Supplier ATCC Supplier No. CRL2397
F98 is a glial-like cell that was isolated from the brain of a rat with undifferentiated malignant glioma. F98 can be used as rat brain tumor models in experimental neuro-oncology.