Learn More
Bachem FA-Ala-Arg-OH
SDP
Catalog No. 50260465
Encompass_Preferred
FA-Ala-Arg-OH, 76079-06-6, C16H23N5O5, Mr 365.39, 50mg. Substrate for human plasma carboxypeptidase N and membrane-bound carboxypeptidase D.
Catalog No. 50-260-465 Supplier Bachem Supplier No. 4013709.0050
FA-Ala-Arg-OH, 76079-06-6, C16H23N5O5, Mr 365.39, 50mg. Substrate for human plasma carboxypeptidase N and membrane-bound carboxypeptidase D.