Learn More
Hach Company Free Ammonia Chlorinating Solution, 4 mL SCDB
Supplier:  Hach Company 2877436
Encompass
Improved reagent for determination of Free Ammonia by the Indophenol method. Replaces 2877336. Requires new procedure (See Reagent/Procedure tab) and instrument software update. See DownloadsOtherProduct Notice-Monochloramine and Free Ammonia Hach Method 10201 4ml self contained dropper bottle (SCDB). Part of reagent set 2879200.
Catalog No. 50-199-6758