Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FA-Gly-Leu-NH2 | 26400-33-9 | 99.5% | 307.34 | 50 MG
SDP

Catalog No. 5000351494
Encompass_Preferred

FA-Gly-Leu-NH2 (FAGLA) is a dipeptide substrate for proteases. It can be hydrolyzed by thermophilic and neutral proteases, exhibiting pH-dependent variations in kcat/Km.

  • Appears as a solid
  • Color ranges from white to light yellow
  • Soluble in DMSO (≥ 350 mg/mL)
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-51494 Supplier Medchemexpress LLC Supplier No. HY13730350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FA-Gly-Leu-NH2 (FAGLA) is a dipeptide substrate for proteases. It can be hydrolyzed by thermophilic and neutral proteases, exhibiting pH-dependent variations in kcat/Km.

  • Appears as a solid
  • Color ranges from white to light yellow
  • Soluble in DMSO (≥ 350 mg/mL)
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.