Learn More
Medchemexpress LLC FA-Leu-Gly-Pro-Ala-OH | 78832-65-2 | 99.4% | 476.52 | 50 MG
SDP
Catalog No. 5000400370
Encompass_Preferred
FA-Leu-Gly-Pro-Ala-OH is a furylacryloyl-terminal tetrapeptide that acts as a substrate for bacterial collagenase and spirochete metalloendopeptidase. It is specifically hydrolyzed at the Leu-Gly bond by spirochete collagenase. This product can be utilized to determine the equilibrium constant of peptide bond hydrolysis and to detect collagenase-mediated cleavage reactions through turbidimetry based on absorbance reduction.
  • Acts as a substrate for bacterial collagenase and spirochete metalloendopeptidase
  • Specifically hydrolyzed at the Leu-Gly bond by spirochete collagenase
  • Utilized to determine the equilibrium constant of peptide bond hydrolysis
  • Detects collagenase-mediated cleavage reactions through turbidimetry based on absorbance reduction
Catalog No. 50-004-00370 Supplier Medchemexpress LLC Supplier No. HYP266150MG
FA-Leu-Gly-Pro-Ala-OH is a furylacryloyl-terminal tetrapeptide that acts as a substrate for bacterial collagenase and spirochete metalloendopeptidase. It is specifically hydrolyzed at the Leu-Gly bond by spirochete collagenase. This product can be utilized to determine the equilibrium constant of peptide bond hydrolysis and to detect collagenase-mediated cleavage reactions through turbidimetry based on absorbance reduction.
  • Acts as a substrate for bacterial collagenase and spirochete metalloendopeptidase
  • Specifically hydrolyzed at the Leu-Gly bond by spirochete collagenase
  • Utilized to determine the equilibrium constant of peptide bond hydrolysis
  • Detects collagenase-mediated cleavage reactions through turbidimetry based on absorbance reduction