Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ Series 315 Laser Embedding Cassettes
Shop All Epredia Products
Click to view available options
Color:
Aqua
Beige
Blue
Gray
Green
Lilac
Ochre
Orange
Pink
Red
White
Yellow
Description
- Epredia™ Laser Cassettes designed for use on both the NOVA™ and VEGA™ Laser Printers
- Contains a special constituent chemical compound that carbonizes when exposed to the laser beam
- Embedding style of cassette comes in different colors
- Required reusable metal cassette lids (Part number FTML315) are sold separately
- Embedding cassette style is designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into Epredia™ cassette hoppers for printing
- Series 315 comes in taped stacks of 40 cassettes; 25 stacks per case
- Epredia™ Laser Cassette Series is not compatible with thermal tape style printers
Specifications
Specifications
Pore Style | Round Holes |
Quantity | 40 Cassettes/Taped Stack, 45 Stacks/Cs. |
Closure Included | No |
Color | White |
Product Type | Cassette |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction