Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Series 315 Laser Embedding Cassettes

Catalog No. 3150318LM Shop All Epredia Products
Click to view available options
Color:
Aqua
Beige
Blue
Gray
Green
Lilac
Ochre
Orange
Pink
Red
White
Yellow
This item is not returnable. View return policy

Catalog No. 3150318LM

Supplier: Epredia™ 3150318LM

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Manufactured specifically for laser printing, the Epredia™ Series 315 Embedding Cassettes have round holes to maximize fluid exchange.

  • Epredia™ Laser Cassettes designed for use on both the NOVA™ and VEGA™ Laser Printers
  • Contains a special constituent chemical compound that carbonizes when exposed to the laser beam
  • Embedding style of cassette comes in different colors
  • Required reusable metal cassette lids (Part number FTML315) are sold separately
  • Embedding cassette style is designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into Epredia™ cassette hoppers for printing
  • Series 315 comes in taped stacks of 40 cassettes; 25 stacks per case
  • Epredia™ Laser Cassette Series is not compatible with thermal tape style printers

Specifications

Pore Style Round Holes
Quantity 40 Cassettes/Taped Stack, 45 Stacks/Cs.
Closure Included No
Color Blue
Product Type Cassette
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.