Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Series 615 Laser Bionet Cassettes with Lids

Catalog No. 6150110LM Shop All Epredia Products
Change view
Click to view available options
Color:
Aqua
Beige
Blue
Gray
Green
Lilac
Ochre
Orange
Pink
Red
White
Yellow
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
6150110LM White
6150210LM Yellow
6150310LM Blue
6150410LM Pink
6150510LM Green
61-506-10LMI Orange
61-507-10LMI Gray
61-508-10LMI Aqua
61-509-10LMI Lilac
61-510-10LMI Beige
61-511-10LMI Ochre
61-512-10LMI Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. 6150110LM Supplier Epredia™ Supplier No. 6150110LMI
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manufactured specifically for laser printing, the Epredia™ Series 615 Bionet Cassettes come with separate lids and are designed for very small and friable tissue samples.

  • Epredia™ Laser Cassettes are designed for use on both the NOVA™ and VEGA™ Laser Printers
  • Contains a special constituent chemical compound that carbonizes when exposed to the laser beam
  • Series 615 comes in multiple different colors with color-matched lids
  • Designed to maximize fluid exchange during processing while still protecting very small and friable tissue samples without the use of sponges or lens paper
  • Come in stacks stripped together by string for ease of loading into Epredia™ cassette hoppers for printing
  • Series 615 comes in stripped stacks of 40 cassettes; 25 stacks per case
  • Epredia™ Laser Cassette is not compatible with thermal tape style printers

Specifications

Pore Style Very Small Squares
Quantity 40 Cassettes/Taped Stack, 25 Stacks/Cs.
Closure Included Yes
Closure Type Separate Lid
Color White
Product Type Cassette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.