Learn More
TARGETMOL CHEMICALS INC FA16 5MG
SDP
Catalog No. 502579370
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. FA16 is a selective metabolically stable ferroptosis inducer with an IC50 value of 1.26 uM.FA16 is a derivative of 2-(trifluoromethyl)benzimidazole.FA16 inhibits the cystine/glutamate inverse transporter protein (system Xc-)-mediated exchange of intracellular glutamate for extracellular cystine.FA16 significantly inhibited tumor growth in a HepG2 xenograft tumor model. HepG2 xenograft tumor model significantly inhibited tumor growth.
Catalog No. 50-257-9370 Supplier TARGETMOL CHEMICALS INC Supplier No. T643575MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. FA16 is a selective metabolically stable ferroptosis inducer with an IC50 value of 1.26 uM.FA16 is a derivative of 2-(trifluoromethyl)benzimidazole.FA16 inhibits the cystine/glutamate inverse transporter protein (system Xc-)-mediated exchange of intracellular glutamate for extracellular cystine.FA16 significantly inhibited tumor growth in a HepG2 xenograft tumor model. HepG2 xenograft tumor model significantly inhibited tumor growth.