Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ FAA Solution
SCI_ED

Catalog No. s25688 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25688 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S25688
This item is not returnable. View return policy

This product is an Educational Grade Chemical — no C of A is available.

Specifications

Chemical Name or Material FAA Solution
CAS 50-00-0, 67-56-1, 7732-18-5, 64-19-7, 64-17-5
Density 0.85 to 0.9g/mL
Packaging Solution
Quantity 500 mL
Assay Percent Range 0.058
Synonym Formalin-Acetic Acid Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.