Learn More
Rockland Immunochemicals Fab Rabbit IgG (H&L) Antibody 1 mg
SDP
Supplier:  Rockland Immunochemicals 8111102
Encompass
Fab Anti-Rabbit IgG (H&L) Antibody generated in goat detects immunoglobulin g from rabbit both heavy and light chains of the antibody molecule are present. Each IgG has two antigen binding sites. Representing approximately 75% of serum immunoglobulins IgG is the most abundant antibody isotype found in the circulation. IgG molecules are synthesized and secreted by plasma B cells. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product consideration must be given to species and immunoglobulin specificity conjugate type fragment and chain specificity level of cross-reactivity and host-species source and fragment composition.
Catalog No. 50-194-3959