Learn More
Advanced Targeting Systems FAB-ZAP HUMAN ANTIBODY INTERNA
Supplier:  Advanced Targeting Systems KIT51Z4
Encompass
Fab-ZAP human Antibody Internalization Kit [KIT-51] 1 kit, tests 4 abs, Host: Goat Reactivity: Human; Conjugate: saporin; Usage: eliminates cells, screen antibodies.
Catalog No. NC1762032