Learn More
Advanced Targeting Systems Fab-ZAP human, IT-51
Catalog No. NC1890034
Encompass
Fab-ZAP human is a chemical conjugate of goat anti-rabbit monovalent antibody and the ribosome-inactivating protein, saporin. It uses your human primary antibody to target and eliminate cells. This secondary conjugate is used to evaluate the potential of a primary antibody to internalize.
Catalog No. NC1890034 Supplier Advanced Targeting Systems Supplier No. IT5125
Fab-ZAP human is a chemical conjugate of goat anti-rabbit monovalent antibody and the ribosome-inactivating protein, saporin. It uses your human primary antibody to target and eliminate cells. This secondary conjugate is used to evaluate the potential of a primary antibody to internalize.