Learn More
Cayman Chemical BMS309403, 10 mg
SDP
Supplier:  Cayman Chemical 1001020610
Encompass_Preferred
A cell-permeable, potent inhibitor of FABP4 (Ki is = 250 and 350 nM, respectively); reduces atherosclerosis in mice lacking ApoE and protects against the development of insulin resistance associated with genetic or diet-induced obesity in mice
Catalog No. NC0688851