Learn More
GENOVIS INC FABALACTICA FAB KIT MIDISPIN
Catalog No. NC1443222
Encompass
Digest and purify up to 10 mg hIgG11 Immobilized FabALACTICA Midispin column + 1 Fc fragment affinity column.FabALACTICA Fab kit for generation and isolation of pure Fab fragments from 1-10 mg human IgG1. The kit consist of 2 spin columns – one for fragmentation (Immobilized FabALACTICA column) and one for purification of Fab fragments,The Immobilized FabALACTICA column is FabALACTICA enzyme immobilized on agarose and the second column has affinity ligands selective for Fc fragments.
This item is not returnable. View return policy
Catalog No. NC1443222 Supplier GENOVIS INC Supplier No. A2AFK100
Digest and purify up to 10 mg hIgG11 Immobilized FabALACTICA Midispin column + 1 Fc fragment affinity column.FabALACTICA Fab kit for generation and isolation of pure Fab fragments from 1-10 mg human IgG1. The kit consist of 2 spin columns – one for fragmentation (Immobilized FabALACTICA column) and one for purification of Fab fragments,The Immobilized FabALACTICA column is FabALACTICA enzyme immobilized on agarose and the second column has affinity ligands selective for Fc fragments.