Learn More
GENOVIS INC Lyophilized FabALACTICA (IgdE), 2000 units
Catalog No. NC1325901
Encompass
Lyophilized FabALACTICA (IgdE) for digestion of 2 mg of human IgG1. FabALACTICA (IgdE) is a unique protease that cleaves human IgG1 at one single amino acid position just above the hinge region, generating intact Fc and Fab fragments
This item is not returnable. View return policy
Catalog No. NC1325901 Supplier GENOVIS INC Supplier No. A0AG1020
Lyophilized FabALACTICA (IgdE) for digestion of 2 mg of human IgG1. FabALACTICA (IgdE) is a unique protease that cleaves human IgG1 at one single amino acid position just above the hinge region, generating intact Fc and Fab fragments