Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fabomotizole hydrochloride | 173352-39-1 | 100.0% | 343.87 g/mol | C15H22ClN3O2S | 10 MG
SDP

Catalog No. 5000285591
Encompass_Preferred

Fabomotizole hydrochloride is the hydrochloride salt of fabomotizole (CM346), an anxiolytic and neuroprotective research compound used in preclinical studies. It is supplied as a high-purity solid for laboratory research into neurological effects and antiparasitic activity.

  • Has anxiolytic and neuroprotective activity observed in preclinical studies.
  • Shows activity against Giardia lamblia and potential antiparasitic effects.
  • High purity: 99.95% (analytical grade).
  • Chemical formula C15H22ClN3O2S; molecular weight 343.87 g/mol.
  • Supplied in small research quantities suitable for assay development.
  • Store sealed at 4°C; in solution, store -80°C for long term or -20°C short term.

Catalog No. 50-002-85591 Supplier Medchemexpress LLC Supplier No. HY14895A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fabomotizole hydrochloride is the hydrochloride salt of fabomotizole (CM346), an anxiolytic and neuroprotective research compound used in preclinical studies. It is supplied as a high-purity solid for laboratory research into neurological effects and antiparasitic activity.

  • Has anxiolytic and neuroprotective activity observed in preclinical studies.
  • Shows activity against Giardia lamblia and potential antiparasitic effects.
  • High purity: 99.95% (analytical grade).
  • Chemical formula C15H22ClN3O2S; molecular weight 343.87 g/mol.
  • Supplied in small research quantities suitable for assay development.
  • Store sealed at 4°C; in solution, store -80°C for long term or -20°C short term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.