Learn More
Medchemexpress LLC Fabomotizole hydrochloride | 173352-39-1 | 99.9% | 343.87 | 100 MG
SDP
Catalog No. 5000361130
Encompass_Preferred
Fabomotizole hydrochloride (CM346) is a compound with anxiolytic and neuroprotective activities. It also shows activity against Giardia lamblia and may inhibit giardiasis.
  • Anxiolytic activities
  • Neuroprotective activities
  • Activity against Giardia lamblia
  • Potential to inhibit giardiasis
  • GABAergic involvement
  • NGF and BDNF release promoting
  • MT1 receptor antagonism
  • MT3 receptor antagonism
  • Sigma agonism
  • Shown to inhibit MAO-A reversibly
  • May involve serotonin receptors
Catalog No. 50-003-61130 Supplier Medchemexpress LLC Supplier No. HY14895A100MG
Fabomotizole hydrochloride (CM346) is a compound with anxiolytic and neuroprotective activities. It also shows activity against Giardia lamblia and may inhibit giardiasis.
  • Anxiolytic activities
  • Neuroprotective activities
  • Activity against Giardia lamblia
  • Potential to inhibit giardiasis
  • GABAergic involvement
  • NGF and BDNF release promoting
  • MT1 receptor antagonism
  • MT3 receptor antagonism
  • Sigma agonism
  • Shown to inhibit MAO-A reversibly
  • May involve serotonin receptors