Learn More
Medchemexpress LLC Fabomotizole hydrochloride | 173352-39-1 | 99.9% | 343.87 | 50 MG
SDP
Catalog No. 5000360824
Encompass_Preferred
Fabomotizole hydrochloride (CM346 hydrochloride) is a compound recognized for its anxiolytic and neuroprotective properties. It also demonstrates activity against *Giardia lamblia*, showing potential to inhibit giardiasis. While its precise mechanism of action is still being elucidated, research suggests possible involvement in GABAergic pathways, promotion of NGF and BDNF release, antagonism of MT1 and MT3 receptors, sigma agonism, reversible inhibition of MAO-A, and potential interaction with serotonin receptors.
  • Anxiolytic activity
  • Neuroprotective activity
  • Activity against Giardia lamblia with potential to inhibit giardiasis
Catalog No. 50-003-60824 Supplier Medchemexpress LLC Supplier No. HY14895A50MG
Fabomotizole hydrochloride (CM346 hydrochloride) is a compound recognized for its anxiolytic and neuroprotective properties. It also demonstrates activity against *Giardia lamblia*, showing potential to inhibit giardiasis. While its precise mechanism of action is still being elucidated, research suggests possible involvement in GABAergic pathways, promotion of NGF and BDNF release, antagonism of MT1 and MT3 receptors, sigma agonism, reversible inhibition of MAO-A, and potential interaction with serotonin receptors.
  • Anxiolytic activity
  • Neuroprotective activity
  • Activity against Giardia lamblia with potential to inhibit giardiasis