Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human FABP-3 ELISA Kit

Catalog No. BMS2263TEN
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BMS2263TEN 10 x 96 Tests
BMS2263 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BMS2263TEN Supplier Invitrogen™ Supplier No. BMS2263TEN
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Human FABP-3 ELISA quantitates Hu FABP-3 in human serum, plasma or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu FABP-3.

Principle of the method

The Human FABP-3 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, binding to the target on a different epitope from the capture antibody. A conjugated enzyme has been incorporated into the assay. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

Rigorous validation

Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Fatty acid binding proteins (FABP) are small (approximately 13-14 kDa) intracellular proteins with a high degree of tissue specificity. FABPs are a class of cytoplasmic proteins that bind long chain fatty acids. They are abundantly present in various cell types and seem to play an important role in the intracellular utilization of fatty acids. There are at least six distinct types of FABP, each showing a specific pattern of tissue expression. FABP leaks, due to its small size, rapidly out of is chemically damaged dying cells leading to a rise in serum levels. Liver FABP is a sensitive marker for cell damage of liver cells in vitro and in vivo. Is chemically damaged tissues are characterized histologically by absence (or low presence) of FABP facilitating recognition of such areas. Next to this L FABP is a marker for rapid hepatocyte lysis in vitro (as for example in toxicology assays) and for detection of liver damage during and after transplantation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05413
Assay Range 156.3-5000 pg/mL
Assay Sensitivity 26.88 pg/mL
Content And Storage Store in refrigerator (2–8°C).
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Growth Factors & Receptors
Sample Type Cell Lysates, Plasma, Serum, Supernatant
Substrate Type Adaptor Proteins
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 422 protein; Cardiac Fatty Acid Binding Protein; FABP 3; FABP11; Fabp3; Fabph1; Fabph-1; Fabph4; Fabph-4; fatty acid binding protein 11; Fatty acid binding protein 11 (FABP11); fatty acid binding protein 3; Fatty acid binding protein 3 heart; fatty acid binding protein 3, muscle and heart; Fatty acid binding protein heart; fatty acid binding protein heart 1; fatty acid binding protein heart 4; fatty acid-binding protein 3; Fatty acid-binding protein, heart; heart FABP; heart fatty acid binding protein; Heart type fatty acid binding protein; heart-type fatty acid-binding protein; H-FABP; M FABP; Mammary derived growth inhibitor; Mammary-derived growth inhibitor; MDGI; M-FABP; muscle and heart (mammary derived growth inhibitor); Muscle fatty acid binding protein; Muscle fatty acid-binding protein; Muscle O-FABP; Mylein protein P2 homolog / adopocyte; O-FABP422 protein; Cardiac Fatty Acid Binding Protein; FABP 3; FABP11; Fabp3; Fabph1; Fabph-1; Fabph4; Fabph-4; fatty acid binding protein 11; Fatty acid binding protein 11 (FABP11); fatty acid binding protein 3; Fatty acid binding protein 3 heart; fatty acid binding protein 3, muscle and heart; Fatty acid binding protein heart; fatty acid binding protein heart 1; fatty acid binding protein heart 4; fatty acid-binding protein 3; Fatty acid-binding protein, heart; heart FABP; heart fatty acid binding protein; Heart type fatty acid binding protein; heart-type fatty acid-binding protein; H-FABP; M FABP; Mammary derived growth inhibitor; Mammary-derived growth inhibitor; MDGI; M-FABP; muscle and heart (mammary derived growth inhibitor); Muscle fatty acid binding protein; Muscle fatty acid-binding protein; Muscle O-FABP; Mylein protein P2 homolog / adopocyte; O-FABP;
Gene ID (Entrez) 2170
Gene Symbol FABP3
Interassay CV 6.2%
Intraassay CV 3.9%
Kit Contents Pre-coated 96 well plate, Standard (2 vials), Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard (2 vials), Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases Fatty acid-binding protein, heart; Fatty acid-binding protein 3; Heart-type fatty acid-binding protein; H-FABP; Mammary-derived growth inhibitor; MDGI; Muscle fatty acid-binding protein; M-FABP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 50 μL, Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet Ice
Storage Requirements 2-8°C
Target FABP3
Test Time 1 hr 20 min
Total Assay Time 4 hr
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.