Learn More
AdipoGen FABP1 human rec His
SDP
Catalog No. 502563047
Encompass_Preferred
Protein. Human FABP1 aa 1-127 is fused at the N-terminus to a His-tag. Source E. coli. Endotoxin content <1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in 20mM Tris-HCl, pH 8. Purity >90% SDS-PAGE. FABP1 is a fatty acid binding protein in adipocytes. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABPs regulate the fatty acid uptake, transport, and metabolism. FABP1 is required for cholesterol synthesis and metabolism.
Catalog No. 50-256-3047 Supplier AdipoGen Supplier No. AG40A0039TC050
Protein. Human FABP1 aa 1-127 is fused at the N-terminus to a His-tag. Source E. coli. Endotoxin content <1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in 20mM Tris-HCl, pH 8. Purity >90% SDS-PAGE. FABP1 is a fatty acid binding protein in adipocytes. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABPs regulate the fatty acid uptake, transport, and metabolism. FABP1 is required for cholesterol synthesis and metabolism.