Learn More
Aviva Systems Biology Corp FABP3 Antibody - N-terminal region (ARP33823_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33823_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against FABP3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human FABP3. Alternate names: MDGI, FABP11, H-FABP, M-FABP, O-FABPApplicationIHC, WB. Species reactivity: Horse, Human, Rabbit, Rat
Catalog No. 50-014-55162