Learn More
Enzo Life Sciences FABP4 inhibitor (5mg). CAS: 300657-03-8
SDP
Catalog No. 502013401
Encompass_Preferred
Alternative name: Adipocyte fatty-acid-binding protein inhibitor, ((2'-(5-Ethyl-3,4-diphenyl-1H-pyrazol-1-yl)(1,1'-biphenyl)-3-yl)oxy)-acetic acid, BMS 309403. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO or ethanol. Long Term Storage: -80°C.
Catalog No. 50-201-3401 Supplier Enzo Life Sciences Supplier No. ENZCHM1180005
Alternative name: Adipocyte fatty-acid-binding protein inhibitor, ((2'-(5-Ethyl-3,4-diphenyl-1H-pyrazol-1-yl)(1,1'-biphenyl)-3-yl)oxy)-acetic acid, BMS 309403. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO or ethanol. Long Term Storage: -80°C.