Learn More
Aviva Systems Biology Corp Fabp5 Antibody - C-terminal region (ARP60048_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60048_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Fabp5. It was validated on Western Blot. . Alternate names: Klbp, mal1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-286-932