Learn More
Medchemexpress LLC FABP5 Human His 100ug
SDP
Catalog No. 5000181413
Encompass_Preferred
FABP5 acts as an intracellular carrier of long-chain fatty acids and related lipids and regulates ligand metabolism It selectively delivers specific fatty acids from the cytoplasm to the nucleus activating nuclear receptors including peroxisome proliferator-activated receptor delta promoting proliferation and survival FABP5 Protein Human (His) is the recombinant human-derived FABP5 protein expressed by E coli with N-6 His labeled tag
Catalog No. 50-001-81413 Supplier Medchemexpress LLC Supplier No. HYP72660100UG
FABP5 acts as an intracellular carrier of long-chain fatty acids and related lipids and regulates ligand metabolism It selectively delivers specific fatty acids from the cytoplasm to the nucleus activating nuclear receptors including peroxisome proliferator-activated receptor delta promoting proliferation and survival FABP5 Protein Human (His) is the recombinant human-derived FABP5 protein expressed by E coli with N-6 His labeled tag