Learn More
ACROBiosystems Recombinant Protein;Human FABP5 / E-FABP Protein;E.coli;1MG;FA5,FABP5,E-FABP,PA-FABP,FABP-5,EFABP,PAFABP
Supplier:  ACROBiosystems FA5H51401MG
Encompass_Preferred
Recombinant Protein;Human FABP5 / E-FABP Protein;1MG;Human FABP5, His Tag (FA5-H5140) is expressed from E.coli cells. It contains AA Ala 2 - Glu 135 (Accession # AAH19385).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 16 kDa. The protein migrates as 16 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8247