Learn More
Aviva Systems Biology Corp FABP9 Antibody - middle region (ARP65665_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65665_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FABP9. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of Human FABP9. Alternate names: PERF, PERF15, T-FABPApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-288-533