Learn More
Fisherbrand™ Fabric ESD Chair - Desk Height with Adjustable Arms and ESD Stationary Glides in ESD Fabric
Catalog No. 14400213 Shop All Fisherbrand Products
Change view
Click to view available options
Desk Height with Adjustable Arms & ESD Stationary Glides in ESD Blue Fabric
Catalog No.
14-400-213 Desk Height with Adjustable Arms & ESD Stationary Glides in ESD Blue Fabric
1 options
Catalog No. 14-400-213
Get 1 Extra Fisherbrand Chair When You Buy 4
View all promotions
Add to Cart
Add to Cart
Fisherbrand™ Fabric ESD Chairs are height adjustable for 31 in. work surfaces and feature ergonomic adjustments, electro-static components such as fabric and stationary glides, and adjustable arms. Chairs are manufactured to ensure proper grounding for static sensitive applications.
  • Adjustable from 19-24 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Electro-static dissipating components to ensure proper grounding
  • Cast-aluminum base
  • Stationary ESD glides suitable for hard floors
  • Electro-static dissipating fabric
  • Meets ANSI/ESD STM12.1-2013
Specifications
Product Type ESD Chair
Autoclavable Not Autoclavable
With Backrest Yes
With Lumbar Support Yes
Arm Style 6-way adjustable (move up and down, in and out, and twist toward or away from the user)
Weight (English) 41 lb.
With Armrest Yes
With Tilt No
Seat Material Polyurethane foam, heavy-duty ESD fabric
Seating Height Desk Height
Footrest No
Seat Tilt N/A
With Casters No
Foot Type Electro-Static Dissipating, non-marring bell glides (suitable for both hard and carpeted floors)
Color Blue
Certifications/Compliance ANSI/ESD-STM-12.1-2006
Adjustable (Height) 19 to 24 in.
Adjustable Yes
Back Style Standard
Base Material Polished Cast Aluminum
Material Fabric
Seating Type Desk Height
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov